Learn More
3M™ Communication Headsets: Mounting Equipment
Catalog No. 191202646 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Com-Tac or Sweet-Tac Headsets
Guide connects boom microphone harness to communication headset cup
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-120-2646 Com-Tac or Sweet-Tac Headsets
19-120-2644 Guide connects boom microphone harness to communication headset cup
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-120-2646 Supplier 3M™ Supplier No. 7000107801
Add to Cart
Add to Cart
Y-Harness for mounting a boom microphone to the right earcup and enabling connection to a PTT adapter for two-way radio communication
  • For left cup microphone placement on ComTac/SwatTac series
  • For ComTac II use FL6AC harness assembly
  • Standard Peltor wiring set-up
  • 3M Catalog# FL6AB
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Com-Tac or Sweet-Tac Headsets
Model FL6AB
Product Type Left side boom harness