Learn More
3M™ CA System Accessory, Breathing Tube Cover
Catalog No. 19021574 Shop All 3M Products
Change view
Click to view available options
Product Type:
Breathing Tube Cover
Catalog No. Product Type
19-021574 Breathing Tube Cover
1 options
Catalog No. 19-021574 Supplier 3M™ Supplier No. 7100077588
  • $63.50
    $58.65 / Case of 10
    Save $4.85
    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
Extend tube life with Versaflo™ S-922 / W-3228-10 Breathing Tube Covers
  • Designed to cover the Versaflo™ BT-Series Breathing Tubes, hardcap assemblies and GVP assemblies
  • 3M Catalog# BT-922/W-3228-10
TRUSTED_SUSTAINABILITY
Specifications
Product Type Breathing Tube Cover
For Use With (Application) Protect the breathing tube and extend the life
For Use With (Equipment) BE-224, GVP-122, GVP-123, L-122 Breathing Tubes
Model BT-922/W-3228-10