Learn More
MSC 3M CONFORM ANTI-SLIP TAPE
Catalog No. 5000802863
Encompass
The Safety-Walk Conformable Tapes and Treads 500 Series is designed for irregular or contoured surfaces in light to heavy shoe-traffic areas. It features a mineral-coated, high friction slip-resistant surface and an aluminum backing that conforms to corners, curves, and irregular surfaces.
  • Mineral-coated, high friction slip-resistant surface
  • Aluminum backing conforms to corners, curves, and irregular surfaces
  • Aggressive, high-performance adhesive ensures a secure bond
  • Resists wrinkling, curling, tearing, shrinking, and lifting
  • Includes Controltac Film Technology for repositioning before final adhesion
  • Features Comply Adhesive Technology for bubble-free application
  • NFSI High Traction Certified
Catalog No. 50-008-02863 Supplier MSC Supplier No. 51557163
The Safety-Walk Conformable Tapes and Treads 500 Series is designed for irregular or contoured surfaces in light to heavy shoe-traffic areas. It features a mineral-coated, high friction slip-resistant surface and an aluminum backing that conforms to corners, curves, and irregular surfaces.
  • Mineral-coated, high friction slip-resistant surface
  • Aluminum backing conforms to corners, curves, and irregular surfaces
  • Aggressive, high-performance adhesive ensures a secure bond
  • Resists wrinkling, curling, tearing, shrinking, and lifting
  • Includes Controltac Film Technology for repositioning before final adhesion
  • Features Comply Adhesive Technology for bubble-free application
  • NFSI High Traction Certified