Learn More
MSC 3M CONFORM ANTI-SLIP TAPE
Catalog No. 5000767396
Encompass
These slip-resistant tapes and treads are engineered for irregular or contoured surfaces with light to heavy shoe-traffic. Featuring a mineral-coated, high-friction surface and an aluminum backing, they conform to various shapes, ensuring enhanced safety and durability in challenging environments.
  • Conforms to irregular surfaces like diamond plating and ladder rungs
  • High-friction, mineral-coated surface for superior slip resistance
  • Resistant to wrinkling, curling, tearing, and lifting
  • Aggressive, high-performance adhesive ensures secure bond
  • NFSI High Traction Certification for proven safety
  • Repositionable during application for precise placement
  • Withstands temperatures up to 79°C (175°F)
Catalog No. 50-007-67396 Supplier MSC Supplier No. 51557114
These slip-resistant tapes and treads are engineered for irregular or contoured surfaces with light to heavy shoe-traffic. Featuring a mineral-coated, high-friction surface and an aluminum backing, they conform to various shapes, ensuring enhanced safety and durability in challenging environments.
  • Conforms to irregular surfaces like diamond plating and ladder rungs
  • High-friction, mineral-coated surface for superior slip resistance
  • Resistant to wrinkling, curling, tearing, and lifting
  • Aggressive, high-performance adhesive ensures secure bond
  • NFSI High Traction Certification for proven safety
  • Repositionable during application for precise placement
  • Withstands temperatures up to 79°C (175°F)