Learn More
Electron Microscopy Sciences Scotch™ Copper Tape, 1/2" wide 18 YD
SDP
Catalog No. 5028522
Encompass_Preferred
The conductive tape is used in scanning electron microscopy specimen mounting. Copper foil tape (Scotch 1181), with conductive adhesive, 3" core and foil thickness: 1.4 mils. Total thickness: 3mils (.076mm), adhesion: 35oz/in , electrical resistance through adhesive: .005ohm/in
Width: 1/2"
Catalog No. 50-285-22 Supplier Electron Microscopy Sciences Supplier No. 77801
The conductive tape is used in scanning electron microscopy specimen mounting. Copper foil tape (Scotch 1181), with conductive adhesive, 3" core and foil thickness: 1.4 mils. Total thickness: 3mils (.076mm), adhesion: 35oz/in , electrical resistance through adhesive: .005ohm/in
Width: 1/2"