Learn More
3M™ Gray Protective Coverall 4570
Catalog No. 19088805 Shop All 3M Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-088-805 3X-Large
19-088-801 Medium
19-088-802 Large
19-088-803 X-Large
19-088-804 2X-Large
19-088-806 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-088-805 Supplier 3M™ Supplier No. 7000089692
Add to Cart
Add to Cart
This item is not returnable. View return policy
Protective Coverall 4570 helps protect against a range of chemical jets and sprays, and has been certified to Type 3/4/5/6 levels of protection.
3M™ Protective Coverall 4570 offers chemical hold out and mechanical strength for workers in environments with chemical exposure. The suit helps protect against hazardous dusts (Type 5), light liquid splashes (Type 6), low pressure liquid sprays (Type 4) and high pressure liquid jets (Type 3). It is also certified to offer protection against radioactive particles (EN 1073-2) and (EN 14126). This suit achieves the highest class performance on all tests in EN 14126 and passes ASTM F1670 and F1671 for helping to protect against biological contaminants.
  • A Versatile Protective Suit for many Applications
  • Protective coverall helps protect against a range of chemical jets and sprays
  • Heavy-duty fabric with a soft anti-static treated inner layer offers protection and convenience
  • Stitched seams are sealed with a 5-layer chemically protective tape
  • Two storm flaps create a double seal for extra protection
  • Large thumb loops and zipper pulls offer convenience while wearing gloves
  • Knit polyester cuffs for user comfort
  • Anti-static, low-linting, microporous
  • Elastic around the head for secure fit
  • Elastic around the face to ensure secure fit
  • Elastic at the calf to provide closure around the top of the footwear for protection against contaminants
  • Elastic around the ankle for protection against contaminants and enhanced fit
  • Universal size with ties to provide a good fit for a wide range of users
  • Lightweight and comfortable to wear
TRUSTED_SUSTAINABILITY
Specifications
Size 3X-Large
Material Polyethylene
Color Gray
Waist Style Elastic
Cuff Style Elastic
Hood Type 3-panel
Closure Type Front Zipper with Double Storm Flap
Seam Type Taped
Protection Class 3
Latex Free Yes
Model 4570-3XL
No. of Pockets None
Size (English) Chest 49 – 52 in.
Size (Metric) Chest 124 to 132 cm
For Use With (Application) Chemical mixing and handling, chemical clean-up, hazardous waste remediation, decontamination, tank cleaning, handling toxic powders, infective agents