Learn More
Enterprise Technology Solutions 3M Deluxe Headgear, w/Ratchet Adj, Plastic, 14"x8", Blue
SDP
Catalog No. 5000828449
Encompass_Preferred
Deluxe headgear helps offer durable head and face protection to workers in metalworking, utilities, woodworking and many other industries. This combination includes H8A ratchet headgear and WP96 polycarbonate faceshield. Clear, molded faceshield provides high impact protection. High-strength thermoplastic crown protector provides impact-resistance. Five-position crown strap and adjustable, 13-position ratchet suspension help provide a precise fit. Headgear requires protective eyewear (sold separately). Recommended applications include facility maintenance, grinding, machine operations, sanding, welding, demolition, painting, blasting, assembly and cleaning.
Catalog No. 50-008-28449 Supplier Enterprise Technology Solutions Supplier No. MMM8250100000
Deluxe headgear helps offer durable head and face protection to workers in metalworking, utilities, woodworking and many other industries. This combination includes H8A ratchet headgear and WP96 polycarbonate faceshield. Clear, molded faceshield provides high impact protection. High-strength thermoplastic crown protector provides impact-resistance. Five-position crown strap and adjustable, 13-position ratchet suspension help provide a precise fit. Headgear requires protective eyewear (sold separately). Recommended applications include facility maintenance, grinding, machine operations, sanding, welding, demolition, painting, blasting, assembly and cleaning.