Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Disposable Overboot Covers 450

Catalog No. 17624137 Shop All 3M Products
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-624-137 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-624-137 Supplier 3M™ Supplier No. 7000088638
Add to Cart
Add to Cart
This item is not returnable. View return policy

The 450 disposable overboot covers from 3M are disposable accessories designed to provide basic barrier protection to help protect footwear against light liquid splashes and hazardous dusts.

3M™ Overboot Covers are a reliable, disposable boot cover designed to help protect footwear against light liquid splashes and hazardous dusts. Offered as an accessory, the overboots are made from a microporous laminate material with an anti-static treatment.

  • Covers are designed with ties and elastic at the calf to help ensure a good fit
  • Distinctive unisex design
  • Integrated sideshields with adjustable lens angle and temple lengths
  • Vertically vented browgard minimizes fogging
  • 3M Catalog# 450

TRUSTED_SUSTAINABILITY

Specifications

Size Universal
Material Microporous Fabric
Color White
Disposable Single-use
Closure Type Elastic Top
Sole Type Non-skid
Model 450
For Use With (Application) Assembly, Cleaning, Machine Operations
Resistance Dry Particulates, Fluid
Product Type Boot Covers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.