Learn More
Enterprise Technology Solutions 3M Double-Coated Tape, 3" Core, 2"x36 Yards, Clear
SDP
Catalog No. 5000828834
Encompass_Preferred
This double-sided tape is made with natural rubber and uses a densified Kraft green plaid paper carrier for increased strength and dimensional stability. It offers excellent high initial adhesion and good holding power to a wide variety of materials. Use tape for indoor and room temperature applications.
Catalog No. 50-008-28834 Supplier Enterprise Technology Solutions Supplier No. MMM410M2X36
This double-sided tape is made with natural rubber and uses a densified Kraft green plaid paper carrier for increased strength and dimensional stability. It offers excellent high initial adhesion and good holding power to a wide variety of materials. Use tape for indoor and room temperature applications.