Learn More
Enterprise Technology Solutions 3M Double-sided Tape,w/Dispenser,Permanent,1/2"x450",CL
SDP
Catalog No. 5000828926
Encompass_Preferred
Double-sided tape is coated with permanent adhesive on both sides of the tape, making it the perfect no-mess alternative to glue for light-duty attaching and mounting tasks. The tape pulls off the roll smoothly and cuts easily so no liner to deal with. Long-aging tape won't dry out or yellow. It is photo-safe (determined in accordance with ISO Standard 18916). Tape comes on a compact handheld dispenser that stores easily in a drawer.
Catalog No. 50-008-28926 Supplier Enterprise Technology Solutions Supplier No. MMM137
Double-sided tape is coated with permanent adhesive on both sides of the tape, making it the perfect no-mess alternative to glue for light-duty attaching and mounting tasks. The tape pulls off the roll smoothly and cuts easily so no liner to deal with. Long-aging tape won't dry out or yellow. It is photo-safe (determined in accordance with ISO Standard 18916). Tape comes on a compact handheld dispenser that stores easily in a drawer.