Learn More
3M™ E-A-Rcaps™ Model 200 Hearing Protector
Catalog No. 173809 Shop All 3M Products
Change view
Click to view available options
Pairs per Package:
10 Pairs/Pk.
Catalog No. Pairs per Package
17-380-9 10 Pairs/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 17-380-9 Supplier 3M™ Supplier No. 7000052716
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ideal for intermittent use, banded hearing protectors are lightweight and feature comfortable foam caps on a flexible band that easily inserts into the ear canal.
  • Band is intended to be worn under the chin
  • Soft foam caps block the entrance to the ear canal
  • Commonly used for visitors or for workers who need intermittent protection
  • One size fits most
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class BL
Size Universal
Headband Material ABS/Polyurethane
For Use With (Application) For Intermittent Worker Protection Needs or for Visitor Use in Noisy Environments
Model 321-2101
Pairs per Package 10 Pairs/Pk.
Plug Material Foam
Plug Color Yellow
Noise Reduction Rating (NRR) 17 dB
Plug Shape Pod
Metal Detectable No
Headband Color Blue