Learn More
3M™ E-A-R™ UltraFit™ 27 Ear Plugs
Catalog No. 19166791 Shop All 3M Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
27 dB
Catalog No. Noise Reduction Rating (NRR)
19-166-791 27 dB
1 options
Catalog No. 19-166-791 Supplier 3M™ Supplier No. 7000127188
Add to Cart
Add to Cart
UltraFit™ reusable ear plugs are durable and feature a triple-flange, push-to-fit design to provide comfort and convenience. Easy to wash with soap and water so they can be used multiple times, reducing waste.
  • Push-to-fit style for easy and hygienic fit
  • Universal size fits comfortably in most ear canals
  • Premolded, triple-flange ear plug design inserts easily
  • No roll-down necessary
  • Soft polymer is washable and reusable
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
Cord Color Clear, blue
Cord Material Vinyl
CSA Z94.2 Class AL
Size Regular
For Use With (Application) Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
High Visibility Yes
Model 340-8002
Packaging Type Poly bag
Pairs per Package 400 Pairs/Cs.
Plug Material Elastomeric polymer
Plug Color Yellow
Noise Reduction Rating (NRR) 27 dB
Corded Corded
Plug Shape Flanged
Product Type Reusable Earplugs
Disposable Reusable
Metal Detectable No
Dielectric No