Learn More
3M™ E-A-Rfit™ Dual-Ear Microphone Assembly
Catalog No. 19122402 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
E-A-Rfit™ Dual-Ear Validation System
Catalog No. For Use With (Equipment)
19-122-402 E-A-Rfit™ Dual-Ear Validation System
1 options
Catalog No. 19-122-402 Supplier 3M™ Supplier No. 7100077867
Add to Cart
Add to Cart
Works in conjunction with the 3M™ E-A-Rfit™ Dual-ear Validation System to help employers determine whether workers are receiving adequate hearing protection from their hearing protector.
  • For use with 3M™ E-A-Rfit™ validation system
  • Tests both ears simultaneously in under five seconds
  • Inserts into back of E-A-Rfit Dual-Ear Enclosure Speaker Assembly
  • Connects to probed test plugs, test pods, or cushions to measure sound pressure levels outside and inside the hearing protector
  • Y-connector between right and left microphone denotes distance user should sit from speaker
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) E-A-Rfit™ Dual-Ear Validation System
Model 073-781
Product Type Dual-Ear, Microphone Assembly