Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ E·A·R·flex™ Hearing Protector Replacement Foam Tips

Catalog No. 189994580 Shop All 3M Products
Change view
Click to view available options
For E-A-Rflex banded hearing protector Model 350-1001; Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-999-4580 For E-A-Rflex banded hearing protector Model 350-1001; Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-999-4580 Supplier 3M™ Supplier No. 7000052865

missing translation for 'validMainframeMsg'

Replacement Foam Tips for 3M™E-A-Rflex™ Hearing Protector (#350-1001).

Replacement Pods for E-A-Rflex™ Hearing Protector 350-1001.
  • Comfortable soft foam
  • Soft foam replacement tip for use only with E-A-Rflex™ banded hearing protector
  • Not for use without band
  • 3M Catalog# 350-1002

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
For Use With (Equipment) For E-A-Rflex™ banded hearing protector
Model 350-1002
Quantity 50/Cs.
Product Type Banded Earplugs
Material Polyurethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.