Learn More
3M™ E-A-Rsoft™ Grippers™ Ear Plugs
Catalog No. 191301918 Shop All 3M Products
Change view
Click to view available options
Ear Plugs; 31 dB; Corded; Large; Bell
Catalog No.
19-130-1918 Ear Plugs; 31 dB; Corded; Large; Bell
1 options
Catalog No. 19-130-1918 Supplier 3M™ Supplier No. 7000127654
Only null left
Add to Cart
Add to Cart
3M™ E-A-Rsoft™ Grippers™ ear plugs unique grip rings help create a soft, noise-blocking seal for effective hearing protection. Ideal for people with larger ear canals.
  • Soft, pliable foam offers comfort for wearing over an extended time
  • Unique grip rings help create a soft, noise-blocking seal
  • Ear plug design offers effective hearing protection for people with larger ear canals
  • Slow recovery foam effectively seals the ear canal and provides reliable hearing protection
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
Cord Color Blue
Cord Material Vinyl
CSA Z94.2 Class AL
Size Large
For Use With (Application) Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
High Visibility Yes
Model 312-6001
Packaging Type Poly bag
Pairs per Package 200 Pairs/Pk.
Plug Material Foam
Plug Color Yellow
Noise Reduction Rating (NRR) 31 dB
Corded Corded
Plug Shape Bell
Disposable Single-use
Metal Detectable No