Learn More
Enterprise Technology Solutions 3M Earplugs, Corded, Push-In, 200PR/BX, Yellow
SDP
Catalog No. 5000828446
Encompass_Preferred
E-A-R Push-Ins corded earplugs provide extended hearing comfort, convenience and protection. One size fits most. Constructed of 3M E-A-Rform foam, these earplugs feature a no roll-down design and a blue stem for a quick, easy fit. Blue stem helps keep the tips clean when the wearer's hands are dirty. Poly bag keeps each pair of earplugs clean before use. Push-to-fit foam is made of polyurethane. Cord is made of vinyl. Noise Reduction Rating (NRR) is 28 decibels CSA Class AL. Corded earplugs are test-compatible with 3M E-A-Rfit Validation System.
Catalog No. 50-008-28446 Supplier Enterprise Technology Solutions Supplier No. MMM3181003
E-A-R Push-Ins corded earplugs provide extended hearing comfort, convenience and protection. One size fits most. Constructed of 3M E-A-Rform foam, these earplugs feature a no roll-down design and a blue stem for a quick, easy fit. Blue stem helps keep the tips clean when the wearer's hands are dirty. Poly bag keeps each pair of earplugs clean before use. Push-to-fit foam is made of polyurethane. Cord is made of vinyl. Noise Reduction Rating (NRR) is 28 decibels CSA Class AL. Corded earplugs are test-compatible with 3M E-A-Rfit Validation System.