Learn More
Enterprise Technology Solutions 3M Earplugs, Uncorded, Classic, 200PR/BX, Yellow
SDP
Catalog No. 5000828460
Encompass_Preferred
E-A-R Classic uncorded earplugs offer a reliable choice in hearing protection and conservation. Slow-recovery, low-pressure foam conforms to the shape of the ear canal to help create a comfortable, snug, noise barrier. These disposable earplugs are flame-resistant and uniquely moisture-resistant. Poly bag keeps PVC earplugs clean before use. Noise Reduction Rating (NRR) is 29 decibels. CSA Class AL. Earplugs are test-compatible with 3M E-A-Rfit Validation System.
Catalog No. 50-008-28460 Supplier Enterprise Technology Solutions Supplier No. MMM3121201
E-A-R Classic uncorded earplugs offer a reliable choice in hearing protection and conservation. Slow-recovery, low-pressure foam conforms to the shape of the ear canal to help create a comfortable, snug, noise barrier. These disposable earplugs are flame-resistant and uniquely moisture-resistant. Poly bag keeps PVC earplugs clean before use. Noise Reduction Rating (NRR) is 29 decibels. CSA Class AL. Earplugs are test-compatible with 3M E-A-Rfit Validation System.