Learn More
Enterprise Technology Solutions 3M Economy Masking Tape, 3" Core, 6RL/PK, 2"x60 Yards
SDP
Catalog No. 5000828831
Encompass_Preferred
Use masking tape for bundling, sealing, holding and other noncritical applications. Tape features a 3" core and a 4.4 mil thickness. Tensile strength is 19 lb. per "
Catalog No. 50-008-28831 Supplier Enterprise Technology Solutions Supplier No. MMM260048A
Use masking tape for bundling, sealing, holding and other noncritical applications. Tape features a 3" core and a 4.4 mil thickness. Tensile strength is 19 lb. per "