Learn More
Enterprise Technology Solutions 3M Electrical Scotch Tape, 10RL/CT, Black
SDP
Catalog No. 5000828712
Encompass_Preferred
Scotch Super 33 Plus Vinyl Electrical Tape offers the highest quality. It is long-lasting with superior adhesion and handling in a wide range of temperatures and conditions. It is designed to perform continuously in a temperature range between 0 to 220 degrees Fahrenheit. Tape provides flexibility and easy handling for all-around performance.
Catalog No. 50-008-28712 Supplier Enterprise Technology Solutions Supplier No. MMM6132BA10
Scotch Super 33 Plus Vinyl Electrical Tape offers the highest quality. It is long-lasting with superior adhesion and handling in a wide range of temperatures and conditions. It is designed to perform continuously in a temperature range between 0 to 220 degrees Fahrenheit. Tape provides flexibility and easy handling for all-around performance.