Learn More
SOLVENTUM 3M Emphaze AEX Series Hybrid Purifier BV8R, Luer, 1/EA
Catalog No. NC1828200
Encompass_Preferred
The Emphaze AEX Hybrid Purifier is an all-synthetic, multi-mechanism clarifying product that contains a Q-functional anion exchange media and a fine particle, bioburden reduction membrane. This product can substantially reduce turbidity, DNA, HCP and bioburden for improved downstream purification.
Catalog No. NC1828200 Supplier SOLVENTUM Supplier No. 70020346873
The Emphaze AEX Hybrid Purifier is an all-synthetic, multi-mechanism clarifying product that contains a Q-functional anion exchange media and a fine particle, bioburden reduction membrane. This product can substantially reduce turbidity, DNA, HCP and bioburden for improved downstream purification.