Learn More
SOLVENTUM 3M™ Emphaze™ AEX Series Hybrid Purifier BV8R | 3in Diameter | 1.8in Length | 0.2 μm
Supplier:  SOLVENTUM 7100158433
Encompass_Preferred
The Emphaze™ AEX Hybrid Purifier is an all-synthetic, multi-mechanism clarifying product that contains a Q-functional anion exchange media and a fine particle, bioburden reduction membrane. This product can substantially reduce turbidity, DNA, HCP and bioburden for improved downstream purification.
Catalog No. NC1805829