Learn More
3M™ Empore™ Vacuum Manifold Accessories, Sealing tape pads
Catalog No. 1437862 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Empore Vacuum Manifold Assembly
Catalog No. For Use With (Equipment)
14-378-62 Empore Vacuum Manifold Assembly
1 options
This item is not returnable. View return policy
Catalog No. 14-378-62 Supplier 3M™ Supplier No. 660
This item is not returnable. View return policy
Specifications
For Use With (Application) Sealing Unused Portions of 96-Well SPE Plates During Vacuum Processing to Enable Uniform Vacuum Application
For Use With (Equipment) Empore Vacuum Manifold Assembly
Product Line Empore
Type Sealing Tape Pad
Material Polypropylene film coated with a synthetic rubber adhesive