Learn More
Enterprise Technology Solutions 3M Eyewear,Anti-Fog,Over-Glass Protection,BK/CL
SDP
Catalog No. 5000828108
Encompass_Preferred
For workers with prescription glasses, OX Protective Eyewear offers an extended field of vision with its wide profile. Seamless lenses with built-in side shields secure an uninterrupted sight range. Eyewear increases comfort and delivers a flexible, polycarbonate construction with over-the-glass coverage. Anti-fog, anti-scratch coating helps minimize fogging. U6-rated lenses absorb up to 99.9 percent of UVA/UVB/UVC rays. Clear design allows the highest visual acuity and color recognition. Temple lengths adjust to fit comfortably. Safety glasses meet Impact-Rated Protector requirements of the ANSI ISEA Z87.1 standard.
Catalog No. 50-008-28108 Supplier Enterprise Technology Solutions Supplier No. MMM1216600000
For workers with prescription glasses, OX Protective Eyewear offers an extended field of vision with its wide profile. Seamless lenses with built-in side shields secure an uninterrupted sight range. Eyewear increases comfort and delivers a flexible, polycarbonate construction with over-the-glass coverage. Anti-fog, anti-scratch coating helps minimize fogging. U6-rated lenses absorb up to 99.9 percent of UVA/UVB/UVC rays. Clear design allows the highest visual acuity and color recognition. Temple lengths adjust to fit comfortably. Safety glasses meet Impact-Rated Protector requirements of the ANSI ISEA Z87.1 standard.