Learn More
MSC 3M FACE SHIELD COVER
Catalog No. 5000792487
Encompass
These standard lens covers are designed for use with Ultimate FX Full Facepiece Reusable Respirators FF-401, 402, and 403. They help extend the service life of the reusable respirator by protecting its lens both during use and when in storage.
  • Replacement part for Ultimate FX Full Facepiece Reusable Respirators FF-401, FF-402, and FF-403
  • Protects the lens of the facepiece
  • Easy to fit over lens
  • Measures 12.7 in overall width x 6.4 in overall height
  • Made of polyester film
  • Intended for industrial/occupational use only
Catalog No. 50-007-92487 Supplier MSC Supplier No. 66688003
These standard lens covers are designed for use with Ultimate FX Full Facepiece Reusable Respirators FF-401, 402, and 403. They help extend the service life of the reusable respirator by protecting its lens both during use and when in storage.
  • Replacement part for Ultimate FX Full Facepiece Reusable Respirators FF-401, FF-402, and FF-403
  • Protects the lens of the facepiece
  • Easy to fit over lens
  • Measures 12.7 in overall width x 6.4 in overall height
  • Made of polyester film
  • Intended for industrial/occupational use only