Learn More
Enterprise Technology Solutions 3M Face Shield,f/X5000 Helmet,High-Impact,CL
SDP
Catalog No. 5000828451
Encompass_Preferred
Face shield attaches to the 3M X5000 Series Helmet (not included). W-Series Face Shield offers optical clarity and high-performance durability in unpredictable environments that involve exposure to impact hazards and flying debris. It quickly mounts to headgear without using tools. Clear polycarbonate provides unrestricted visibility. Splash-resistant, chemical-resistant material meets ANSI Z87 plus for impact-resistance. It resists heat up to 266 degrees Fahrenheit. Contoured shape is comfortable to wear. Durable face shield is recommended for industries such as automotive, design/construction, facility sanitation, food-processing, general manufacturing, heavy infrastructure, industrial maintenance, military maintenance, repair/operation, mining/oil/gas, pharmaceutical and transportation.
Catalog No. 50-008-28451 Supplier Enterprise Technology Solutions Supplier No. MMMWP96
Face shield attaches to the 3M X5000 Series Helmet (not included). W-Series Face Shield offers optical clarity and high-performance durability in unpredictable environments that involve exposure to impact hazards and flying debris. It quickly mounts to headgear without using tools. Clear polycarbonate provides unrestricted visibility. Splash-resistant, chemical-resistant material meets ANSI Z87 plus for impact-resistance. It resists heat up to 266 degrees Fahrenheit. Contoured shape is comfortable to wear. Durable face shield is recommended for industries such as automotive, design/construction, facility sanitation, food-processing, general manufacturing, heavy infrastructure, industrial maintenance, military maintenance, repair/operation, mining/oil/gas, pharmaceutical and transportation.