Learn More
3M™ Fectoggles™ Eyewear
Catalog No. 19003339A Shop All 3M Products
Change view
Click to view available options
Includes:
Elastic Band Frame
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Size
19-003-339A Elastic Band Frame Medium
19-003-339B Elastic Band Frame Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.
Catalog No. 19-003-339A Supplier 3M™ Supplier No. 7010045742
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.
Performance of an impact goggle with the comfort of a spectacle
  • Polycarbonate lens absorbs 99.9% UV
  • DX™ antifog hardcoat protects lens from fog, scratching and most chemicals
  • Unique lens has no parting lines to obstruct or distract wearer's view
  • Lightweight, effective eye protection for high-particulate areas
  • Contoured to conform to the shape of the face to optimize coverage
  • Foam flange seals the entire perimeter of the lens to keep out fine particulates and control sweat buildup
  • Fire-retardant, braided, elastic headband for snug fit
  • 3M Catalog# 16420-00000-10, 16408-00000-10, 16400-00000-10, 16412-00000-10
Compliance
Meets ANSI™ Z87.1-2010 and CSA Z94.3-2007 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2010-High Impact; CSA Z94.3-2007 standards
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Safety goggles
Includes Elastic Band Frame
Size Medium
Material Polycarbonate