Learn More
3M™ Fectoids™ Eyewear
Catalog No. 189992744 Shop All 3M Products
Change view
Click to view available options
Lens Tint: Gray; Frame Color: Blue
Catalog No.
18-999-2744 Lens Tint: Gray; Frame Color: Blue
1 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulatinos and the user instructions, warnings, and limitations according to each product.
Catalog No. 18-999-2744 Supplier 3M™ Supplier No. 16205
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulatinos and the user instructions, warnings, and limitations according to each product.
Adjustable lens angle
  • Sports styling closely follows contours of the face to provide complete coverage
  • DX™ anti-fog hard coat protects lens from fog, scratching, and most chemicals
  • Lenses absorb 99.9% UV
  • Lens angle can be adjusted to one of three positions
  • Length of the temples can be adjusted to four settings for a custom fit
  • Lenses come in two sizes and can be changed quickly and easily without tools
Compliance
Meet ANSI Z87.1-2003 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meet ANSI Z87.1-2003 standards.
Lens Tint Gray
Frame Color Blue
Lens Material Polycarbonate