Learn More
Enterprise Technology Solutions 3M Flatback Tape, 1/2"x 60 Yards, 3" Core, White
SDP
Catalog No. 5000828946
Encompass_Preferred
Made with natural rubber, pressure-sensitive adhesive, this tape bonds to a wide variety of paper, film, fabric and yarn surfaces. It is printable with standard flexographic inks and is also easily marked with ballpoint pens, felt-tip pens, pencils and rubber stamps. The thick, flatback paper backing creates good paint lines when used for masking purposes. Use also for banding items where color-coding is required, last lap end-roll tabbing of paper, film, fabric and yarns and light to medium bundling applications. The tape detaches easily and provides clean removal from a variety of substrates.
Catalog No. 50-008-28946 Supplier Enterprise Technology Solutions Supplier No. MMM25612
Made with natural rubber, pressure-sensitive adhesive, this tape bonds to a wide variety of paper, film, fabric and yarn surfaces. It is printable with standard flexographic inks and is also easily marked with ballpoint pens, felt-tip pens, pencils and rubber stamps. The thick, flatback paper backing creates good paint lines when used for masking purposes. Use also for banding items where color-coding is required, last lap end-roll tabbing of paper, film, fabric and yarns and light to medium bundling applications. The tape detaches easily and provides clean removal from a variety of substrates.