Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Foam Ear Plugs

Catalog No. p-91250 Shop All 3M Products
Change view
Click to view available options
Model:
1100
1110
Corded:
Corded
Uncorded
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Corded
19-072076 1100 Uncorded
19-072077 1110 Corded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-072076 Supplier 3M™ Supplier No. 7100099847
Only null left
Add to Cart
Edge
Add to Cart

Foam ear plugs are an economical choice for disposable hearing protection. Hypoallergenic and a tapered design help provide a noise-reducing seal in the ear canal.

  • Disposable orange ear plugs
  • Made from hypoallergenic material for maximum protection and comfort
  • Smooth, dirt-resistant surface for hygiene
  • Recommended for high noise situations (up to 100 dBA TWA)

Compliance

3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
High Visibility Yes
Model 1100
Packaging Type Poly bag
Pairs per Package 1000 Pairs/Cs
Plug Material Foam
Plug Color Orange
Noise Reduction Rating (NRR) 29 dB
Corded Uncorded
Plug Shape Bullet
Disposable Single-use
Metal Detectable No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.