Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ FR-7800B Respirator Full Facepieces

Catalog No. 191304853C Shop All 3M Products
Change view
Click to view available options
Format:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
19-130-4853C Large
19-130-4853A Small
19-130-4853B Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-130-4853C Supplier 3M™ Supplier No. 7000126540
  • $968.00
    $697.65 / Case of 1

    Save $270.35

    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Integral speaking diaphragm improves voice amplification and clarity

  • Features a unique double flange face sealing surface that enhances fit and protection
  • Designed for first responder applications commonly found in law enforcement, fire, emergency response and medical environments
  • Canister can be mounted on either side to accommodate different weapon sighting styles
  • Lens has optical correction promoting no left-to-right visual distortion
  • 3M Catalog# FR-7800B-L , 3M Catalog# FR-7800B-M , 3M Catalog# FR-7800B-S

Certifications

NIOSH approved to Chemical Biological Radiological Nuclear (CBRN) requirements when used with the 3M Canister FR-15-CBRN (19-130-0673).

Compliance

Meet ANSI™ Z87.1-2003 standards.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH, CBRN, ANSI Z87.1-2003
Material Silicone, Butyl Rubber Faceshield
Product Type Full Facepiece
Format Large
For Use With (Application) CBRN applications when used with 3M™ canister fr-15-CBRN
For Use With (Equipment) Air purifying respirator
Includes Integral speaking diaphragm
Model FR-7800B-L
Suspension Type 6-point strap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.