Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Gel Mousepad Wristrest

Catalog No. 19021853 Shop All 3M Products
Change view
Click to view available options
Color:
Beach
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-021-853 Beach
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-021-853 Supplier 3M™ Supplier No. 7010290915

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

  • 3M Catalog# MW308BH

TRUSTED_SUSTAINABILITY

Specifications

Color Beach
Product Type Gel Mouse Pad Wrist Rest
Length (Metric) 21.8 cm
Length (English) 8.6 in.
Size Compact
For Use With (Application) Inhibits the growth of microorganisms such as bacteria and encourages neutral wrist posture
Includes Non-skid base, clear gel
Material Foam
Model MW308BH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.