Learn More
Uline 3M GoggleGear™ 500 Safety Goggles - Clear Lens
Catalog No. NC2197368
Encompass
This item must be ordered in quantities of 2 per supplier requirements.
Low-profile design fits comfortably underneath respirators and face shields.Protects against impact and chemical splash. Indirect vent.99.9% UV protection.
This item is not returnable. View return policy
Catalog No. NC2197368 Supplier Uline Supplier No. S24392C
This item must be ordered in quantities of 2 per supplier requirements.
Low-profile design fits comfortably underneath respirators and face shields.Protects against impact and chemical splash. Indirect vent.99.9% UV protection.