Learn More
Enterprise Technology Solutions 3M Goggles, Anti-Fog, Adjustable Band, 10/CT, Clear Lens, Gray
SDP
Catalog No. 5000828180
Encompass_Preferred
Safety goggles feature a durable, anti-fog coating that's bonded to the lens to resist fogging for longer, even after washing multiple times. Advanced Scotchgard coating produces a reduced-contact angle, flattening the water beads into a thin, transparent film of water to allow light to pass through and ensure you maintain a clear vision of your surroundings. Plus, it withstands disinfection with diluted bleach soaking or alcohol wipes without losing its anti-fog performance. Safety goggles with indirect ventilation are perfect for working in hot/humid conditions, indoor/outdoor work, physically demanding tasks, climate-controlled areas and more.
Catalog No. 50-008-28180 Supplier Enterprise Technology Solutions Supplier No. MMMGG501SGAFCT
Safety goggles feature a durable, anti-fog coating that's bonded to the lens to resist fogging for longer, even after washing multiple times. Advanced Scotchgard coating produces a reduced-contact angle, flattening the water beads into a thin, transparent film of water to allow light to pass through and ensure you maintain a clear vision of your surroundings. Plus, it withstands disinfection with diluted bleach soaking or alcohol wipes without losing its anti-fog performance. Safety goggles with indirect ventilation are perfect for working in hot/humid conditions, indoor/outdoor work, physically demanding tasks, climate-controlled areas and more.