3M Safety-Walk Slip-Resistant General Purpose Tape 610, part of the 600 Series, is engineered for light to heavy shoe traffic areas, offering a durable, mineral-coated, high-friction surface and a low-profile design to reduce trip hazards.
- High-performance pressure-sensitive adhesive for easy application.
- Ideal for flat surfaces, steps, stairways, entrances, ramps, and ladders.
- NFSI High Traction Certified for superior slip resistance.
- Proprietary Controltac™ Film Technology enables repositioning during application.
- Comply™ Adhesive Technology ensures bubble-free application.
- Features PSA adhesive, mineral anti-slip material, and Poly Coated Paper/Polyester backing.
- Heavy Duty Tread Grade; minimum operating temperature: 4 °C (40 °F).
- Meets ANSI/NFSI B101.1, B101.3, and U.S. MIL-PRF-24667C specifications.