Learn More
3M™ H-700 Series Cap Style Hard Hats with 4-Point Ratchet Suspension
Catalog No. 17623795 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Gray
Green
Navy Blue
Pink
Red
Tan
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-795 Pink
17-623-314 Gray
17-623-321 Navy Blue
17-623-325 Red
17-623-326 Green
17-623-327 Yellow
17-623-785 Tan
17-623-790 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-623-795 Supplier 3M™ Supplier No. 7010385057
Protect top of the head due to potential hazard of small falling objects or exposed fixed objects
  • Suspension adjusts easily to allow for customized height and fit
  • Replaceable brow pad for more comfort and extended shelf life
  • Short brim for upward visibility
  • Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C
  • ANSI/ISEA compliant when worn in the forward and reverse donning positions
  • Available in various colors
  • 3M Catalog# H-701R, H-702R, H-703R, H-704R, H-705R, H-706R, H-707R, H-708R, H-709R, H-710R, H-711R, H-712R, H-713R
TRUSTED_SUSTAINABILITY
Specifications
Color Pink
Model H-713R
Product Type Cap-style Hard Hat