Learn More
3M™ H-700 Series Cap Style Vented Hard Hats
Catalog No. 17623797 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Blue
Green
Pink
Tan
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-797 Pink
17-623-318 Blue
17-623-317 Green
17-623-787 Tan
17-623-792 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-623-797 Supplier 3M™ Supplier No. 7100162727
Helps protect top of the head due to potential hazard of small falling objects or exposed fixed objects
  • Suspension allows to customize height and fit
  • Vents allow air circulation to keep worker cooler and more comfortable
  • Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C
  • ANSI/ISEA compliant when worn in the forward and reverse donning positions
  • Replaceable brow pad for comfort and extends service life
  • Short brim for upward visibility
  • Available in various colors
  • 3M Catalog# H-701V, H-702V, H-703V, H-704V, H-711V, H-712V, H-713V
TRUSTED_SUSTAINABILITY
Specifications
Color Pink
Model H-713V
Product Type Cap-style Hard Hat
Ventilated Ventilated