Learn More
3M™ H-800 Series Full Brim Hard Hats with Uvicator™ Sensors
Catalog No. 17623777 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Blue
Hi-vis Orange
Hi-vis Yellow
Navy Blue
Orange
Pink
Red
Tan
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-777 Pink
17-623-729 Yellow
17-623-730 Blue
17-623-732 Red
17-623-733 Orange
17-623-734 Hi-vis Orange
17-623-737 Navy Blue
17-623-736 Hi-vis Yellow
17-623-738 Tan
17-623-773 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 17-623-777 Supplier 3M™ Supplier No. 7010296269
With Uvicator sensor that changes color as hard hat is exposed to UV light
  • Made of HDPE
  • Features 4-point ratchet suspension with three position height adjustment allows for customized fit
  • Perforated absorbent brow pad 
  • Optional 6-point suspension strap for more comfort
  • Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C, G and E
  • Available in several colors
  • 3M Catalog# H-801R-UV, H-802R-UV, H-803R-UV, H-804R-UV, H-805R-UV, H-806R-UV, H-807R-UV, H-808R-UV, H-809R-UV, H-810R-UV, H-811R-UV, H-812R-UV, H-813R-UV
TRUSTED_SUSTAINABILITY
Specifications
Color Pink
Material HDPE
Model H-813R-UV
Quantity 20/Cs.
Weight (English) 416 oz.
Weight (Metric) 11.79 kg
Head Style Full Brim