Learn More
3M™ H-800 Series Full Brim Vented Hard Hats
Catalog No. 17623778 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Green
Hi-vis Orange
Hi-vis Yellow
Navy Blue
Orange
Pink
Red
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-778 Pink
17-623-718 Yellow
17-623-720 Green
17-623-723 Hi-vis Orange
17-623-722 Orange
17-623-721 Red
17-623-725 Hi-vis Yellow
17-623-726 Navy Blue
17-623-774 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-623-778 Supplier 3M™ Supplier No. 7100162734
3M™ H-800V-UV Series Full Brim Vented Hard Hats with Ratchet Suspension Systems are available with Uvicator™ Sensor. Comfortable, tough and lightweight protective headgear that helps protect the head from small falling objects and fixed objects that may be bumped into Uvicator sensor changes color as the hard hat is exposed to UV light. The sun’s UV rays can cause a hard hat shell to become brittle and compromise its ability to protect the wearer. With the Uvicator sensor, the hard hat should be replaced when the sun smart sensor changes from red to white.
  • Vents built into the hard hat release heat buildup and allow air circulation to enhance comfort of the worker
  • Cooling effect of air circulation helps reduce perspiration in hot and humid working environments
  • 4-point suspension system has unique features that allow for a customized height and fit that best suits the individual needs of the wearer
  • An optional 6-point suspension strap can be added for additional comfort and fit
  • Comfortable suspension sits lower on the head to reduce pressure points and increase security, especially when looking up and bending down
  • Smooth, easy-turning ratchet suspension allows for simple adjustments
  • 3-level height adjustment allows workers to customize the fit
  • Perforated, absorbent brow pad provides enhanced comfort
  • Besides head protection, full brim hard hats offer extra shielding from the sun, splashes, rain, sleet, and snow
  • Full brim hard hats are often preferred in hot and sunny locations because of the extra shading they provide
  • Hardhats are commonly worn in these industries: heavy and light industrial, construction, oil and gas, road work, utilities, forestry, and mining
  • 3M Catalog# H-801V, H-802V, H-803V, H-804V, H-805V, H-806V, H-807V, H-808V, H-809V, H-810V, H-811V, H-812V, H-813V
Meets requirements of ANSI/ISEA Z89.1-2014 Type 1, Class CHigh visibility orange and high visibility yellow colored hard hats meet ANSI Z89.1-2014 High Visibility Performance requirements.
TRUSTED_SUSTAINABILITY
Specifications
Color Pink
Material HDPE
Model H-813V
Quantity 20/Cs.
Size (Metric) 168 to 197 mm
Head Style Full Brim