Learn More
3M™ H-800 Series Full Brim Vented Hard Hats with Uvicator™ Sensors
Catalog No. 17623779 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Blue
Green
Hi-vis Orange
Hi-vis Yellow
Navy Blue
Orange
Pink
Red
Tan
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-779 Pink
17-623-742 Green
17-623-740 Yellow
17-623-741 Blue
17-623-743 Red
17-623-744 Orange
17-623-745 Hi-vis Orange
17-623-747 Hi-vis Yellow
17-623-749 Tan
17-623-748 Navy Blue
17-623-775 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 17-623-779 Supplier 3M™ Supplier No. 7100162737
With Uvicator sensor that changes color as hard hat is exposed to UV light
  • Made of HDPE
  • Features 4-point ratchet suspension with three position height adjustment allows for customized fit
  • Perforated absorbent brow pad 
  • Optional 6-point suspension strap for more comfort
  • Vents allow air circulation to keep worker cooler and more comfortable
  • Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C
  • Available in several colors
  • 3M Catalog# H-801V-UV, H-802V-UV, H-803V-UV, H-804V-UV, H-805V-UV, H-806V-UV, H-807V-UV, H-808V-UV, H-809V-UV, H-810V-UV, H-811V-UV, H-812V-UV, H-813V-UV
TRUSTED_SUSTAINABILITY
Specifications
Color Pink
Model H-813V-UV
Size (Metric) 168 to 197 mm
Suspension Type 4-point Ratchet
Ventilated Ventilated
Head Style Full Brim