Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ H-Series Hood Accessory, Cap Suspension Assembly

Catalog No. 19150672 Shop All 3M Products
Change view
Click to view available options
Cap suspension
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-150-672 Cap suspension
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-150-672 Supplier 3M™ Supplier No. 7000126541

missing translation for 'validMainframeMsg'

For use with H-410-10 and H-420-10 hoods.

  • Provides support for breathing tube connected to rear of hood
  • 3M Catalog# H-213-2

These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH Approved
Color Gray
Disposable Reusable
Product Type Cap Suspension Assembly
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Machine Operation, Painting, Sanding, Welding
For Use With (Equipment) H-Series Hoods H-410-10 and H-420-10
Model H-213-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.