Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ H-Series Hood Assemblies Replacement Hoods

Catalog No. 190881288 Shop All 3M Products
Change view
Click to view available options
Hood replacement
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-088-1288 Hood replacement
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-088-1288 Supplier 3M™ Supplier No. 7000002070
This item is not returnable. View return policy

Provides head and neck protection from pesticide and chemical overspray. H-Series hoods are for use with belt-mounted PAPRs or supplied air components.

  • Lightweight, loose fitting, hood
  • Replacement for 3M™ Hood Assemblies H-411 and H-412
  • 3M Catalog# H-410-10
TRUSTED_SUSTAINABILITY

Specifications

Assigned Protection Factor (APF) 1,000
Color White
Product Type Respirator Hood
For Use With (Application) Assembly, machine operation
For Use With (Equipment) Belt mounted powered air purifying respirator or supplied air component
Includes Collar
Model H-410-10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.