Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ H-Series White Hood Assemblies

Catalog No. 190881269 Shop All 3M Products
Change view
Click to view available options
Hood Assembly W/collar and Hardhat
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-088-1269 Hood Assembly W/collar and Hardhat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-088-1269 Supplier 3M™ Supplier No. 7000002072

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with 3M™ belt-mounted powered air purifying respirators (PAPR) and supplied air respirator (SAR) systems.

  • Feature lightweight Tychem™ QC Fabric
  • Comfortable headgear
  • Can be worn with facial hair and glasses
  • White

3M H-400 Series Hoods

  • Feature lightweight Tychem™ QC Fabric and provide high levels of airflow for maximum comfort for pharmaceutical, laboratory, and health care applications as well as during painting and sanding operations H-400 is available with hard hat and choice of inner shroud or collar
  • 3M Catalog# H-412

These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Assigned Protection Factor (APF) 1,000
Certifications/Compliance NIOSH
Color White
Material Tychem QC
Product Type Hood Assembly
For Use With (Application) Machine operation, military maintenance, repair and operation, pharmaceutical
For Use With (Equipment) Belt mounted powered air purifying respirator and supplied air respirator system
Includes Collar and hardhat
Model H-412
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.