Learn More
3M™ Hard Hat with Uvicator™ Sensor
Catalog No. 17623798 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Blue
Green
Pink
Tan
Ventilated:
Not Ventilated
Ventilated
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Ventilated
17-623-798 Pink Ventilated
17-623-639 Blue Ventilated
17-623-640 Green Not Ventilated
17-623-641 Green Ventilated
17-623-786 Tan Not Ventilated
17-623-788 Tan Ventilated
17-623-793 Black Ventilated
17-623-796 Pink Not Ventilated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-623-798 Supplier 3M™ Supplier No. 7100162730
Protective hard hat with vents, comfortable 4-point ratchet suspension and standard brow pad
Available in a variety of colors, vented or unvented
  • Cap style hard hats constructed of HDPE
  • Four-point ratchet suspension
  • Uvicator lets wearer known when to replace hard hat due to UV exposure
  • Suspension sits lower on head to reduce pressure and increase security
  • Smooth, easy turning ratchet suspension
  • Dielectric: Class C, G, and E
  • 3M Catalog# H-701R-UV, H-701V-UV, H-702R-UV, H-702V-UV, H-703R-UV, H-703V-UV, H-704R-UV, H-704V-UV, H-709R-UV, H-711R-UV, H-711V-UV, H-712V-UV, H-713R-UV, H-713V-UV, H-712R-UV
Compliance
Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C.
TRUSTED_SUSTAINABILITY
Specifications
Color Pink
Model H-713V-UV
Product Type Hard Hat with Uvicator
Suspension Type 4-Point Ratchet
Ventilated Ventilated