Learn More
3M™ Hatshells
Catalog No. 19021671 Shop All 3M Products
Change view
Click to view available options
Type 1; Class E head protection; 5/Cs.
Catalog No.
19-021671 Type 1; Class E head protection; 5/Cs.
1 options
This item is not returnable. View return policy
Catalog No. 19-021671 Supplier 3M™ Supplier No. 7000131000
This item is not returnable. View return policy
When used with the 3M™ Head Suspension W-2878, the 3M™ Hatshell W-3258 meets the requirements of ANSI standard Z89.1-2003, Type 1, Class E head protection.
  • 3M Catalog# W-3258-5
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z89.1-2003
For Use With (Equipment) Head Suspension W-2878
For Use With (Application) Cleaning, electrical, facility maintenance, painting, sanding, welding
Model W-3258-5
Product Type Hatshell