Learn More
MSC 3M Heat Shrink Tubing, Semi-Rigid Polyolefin, 0.5 in, 48 in
Catalog No. 5000507238
Encompass
This heat shrink tubing is a semi-rigid, heat-shrinkable polyolefin product with a 2.5:1 shrink ratio. It features a dual-wall construction, achieved through co-extrusion and selective cross-linking, resulting in an integral, meltable inner wall for effective sealing. The tubing is supplied in 48 inch lengths and packaged with 12 pieces per case.
  • Semi-rigid polyolefin material
  • Dual wall construction with meltable inner wall
  • 2.5:1 shrink ratio
  • Supplied in 48 inch lengths
  • Packaged as 12 pieces per case
  • UL Recognized component
Catalog No. 50-005-07238 Supplier MSC Supplier No. 77677425
This heat shrink tubing is a semi-rigid, heat-shrinkable polyolefin product with a 2.5:1 shrink ratio. It features a dual-wall construction, achieved through co-extrusion and selective cross-linking, resulting in an integral, meltable inner wall for effective sealing. The tubing is supplied in 48 inch lengths and packaged with 12 pieces per case.
  • Semi-rigid polyolefin material
  • Dual wall construction with meltable inner wall
  • 2.5:1 shrink ratio
  • Supplied in 48 inch lengths
  • Packaged as 12 pieces per case
  • UL Recognized component