Learn More
3M™ Half Facepiece Respirator 6000 Series Accessory, Exhalation Valve
Catalog No. 189994560 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Half Facepiece Respirator 6000 Series
Catalog No. For Use With (Equipment)
18-999-4560 Half Facepiece Respirator 6000 Series
1 options
Catalog No. 18-999-4560 Supplier 3M™ Supplier No. 7000002057
Add to Cart
Add to Cart
For use with 6000 Series half facepiece respirators.
  • Recommended for 3M Half Facepiece Respirator 6000 Series (sold separately)
  • Valve maintains original service performance of facepiece
  • 3M Catalog# 6889
TRUSTED_SUSTAINABILITY
Specifications
Product Type Exhalation Valve
For Use With (Application) Assembly, blasting, cleaning, demolition, electrical, facility maintenance, grinding, health and safety testing, machine operation, painting, remediation, sanding, welding
For Use With (Equipment) Half Facepiece Respirator 6000 Series
Model 6889