Learn More
Enterprise Technology Solutions 3M Laminating Pouch, Bus Card, 3-3/4"x2-3/8", 20/PK, CL
SDP
Catalog No. 5000740285
Encompass_Preferred
Laminating pouches are designed for use with the 3M Scotch Thermal Laminator or any standard, thermal-pouch laminator. Pouches provide instant, two-sided protection for business cards or other frequently handled or referred to items. Photo-safe pouches are clear to let important information show through with a professional finish.
Catalog No. 50-007-40285 Supplier Enterprise Technology Solutions Supplier No. MMMTP585120
Laminating pouches are designed for use with the 3M Scotch Thermal Laminator or any standard, thermal-pouch laminator. Pouches provide instant, two-sided protection for business cards or other frequently handled or referred to items. Photo-safe pouches are clear to let important information show through with a professional finish.