Learn More
Andwin Scientific 3M LEAD FREE AUTOCLAVE TAPE 1"
SDP
Catalog No. NC0802040
Encompass
The ESPE™ autoclave steam indicator tape can be used on all wraps, including disposable, non-woven, and linen wraps. This tape is lead free and thus does not require hazardous waste disposal. The stripes change color when exposed to steam sterilization, which then conveniently allows the user to identify the processed items.
Catalog No. NC0802040 Supplier Andwin Scientific Supplier No. 1322C
Add to Cart
Add to Cart
The ESPE™ autoclave steam indicator tape can be used on all wraps, including disposable, non-woven, and linen wraps. This tape is lead free and thus does not require hazardous waste disposal. The stripes change color when exposed to steam sterilization, which then conveniently allows the user to identify the processed items.