Learn More
3M™ High Fluid Resistant Procedure Mask, Light Green
Catalog No. 19177860 Shop All 3M Products
Change view
Click to view available options
High Fluid Resistant Procedure Mask
Catalog No.
19-177-860 High Fluid Resistant Procedure Mask
1 options
Catalog No. 19-177-860 Supplier 3M™ Supplier No. 7000002919
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
3M High Fluid Resistant Procedure Mask with Face Shield is a highly fluid-resistant procedure mask with ear loops. These masks are appropriate for procedures where there is concern about potential exposure to blood and body fluids.
  • Able to warm at higher flow rates
  • Lightweight
  • Comfortable and easy to breath through
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Intended for general patient care and procedure environments, ER, cardiac labs, etc. where high levels of fluid protection is required.
Model 1840