Learn More
3M™ Lexa™ Splash GoggleGear™ Safety Goggles
Catalog No. p-94847 Shop All 3M Products
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-053-614 Medium
19-053-613 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-053-614 Supplier 3M™ Supplier No. 7010045731
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Lightweight, contoured, low-profile design
  • Three lens angle adjustments for custom fit
  • Vent channels at top and bottom designed to help minimize fogging
  • DX™ coating resists fogging, scratching, static and many chemicals
  • 3M Catalog# 16644-00000-10 , 3M Catalog# 16645-00000-10
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2010 and CSA Z94.3-2010 standards
Description Clear ellipsoidal Lexa
Lens Tint Clear
Product Type Safety goggles
Model 16644-00000-10
Size Medium
Ventilation Details Vent channels at top and bottom